Learn More
LEPRE1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human LEPRE1 protein.
Brand: Abnova H00064175-B01P.50ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an enzyme that is a member of the collagen prolyl hydroxylase family. These enzymes are localized to the endoplasmic reticulum and their activity is required for proper collagen synthesis and assembly. Mutations in this gene are associated with osteogenesis imperfecta type VIII. Two alternatively spliced transcript variants encoding different isoforms have been described. Other variants may exist, but their biological validity has not been determined. [provided by RefSeq
Sequence: MLGEEHTRSIGPRESAKEYRQRSLLEKELLFFAYDVFGIPFVDPDSWTPEEVIPKRLQEKQKSERETAVRISQEIGNLMKEIETLVEEKTKESLDVSRLTREGGPLLYEGISLTMNSKLLNGSQRVVMDGVISDHECQELQRLTNVAATSGDGYRGQTSPHTPNEKFYGVTVFKALKLGQEGKVPLQSAHLYYNVTEKVRRIMESYFRLDTPLYFSYSHLVCRTAIEEVQAERKDDSHPVHVDNCILNAETLVCVKEPPAYTFRDYSAILYLNGDFDGGNFYFTELDAKTVTAEVQPQCGRAVGFSSGTENPHGVKAVTRGQRCAIALWFTLDPRHSERDRVQADDLVKMLFSPEEMDLSQEQPLDAQQGPPEPAQESLSGSESKPKDELSpecifications
LEPRE1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human LEPRE1 protein. | |
LEPRE1 | |
GROS1/MGC117314/P3H1 | |
Mouse | |
Affinity chromatography | |
RUO | |
64175 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC015309.1 | |
LEPRE1 | |
LEPRE1 (AAH15309, 1 a.a. ∼ 390 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |