Learn More
Legumain/Asparaginyl Endopeptidase Mouse anti-Human, Clone: 312109, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB21992
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469255
  • £417.48 / 500µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Legumain/Asparaginyl Endopeptidase Monoclonal antibody MAB21992 Specifically detects Legumain/Asparaginyl Endopeptidase in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
Legumain/Asparaginyl Endopeptidase
312109
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q99538
AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects both the pro and mature forms of human Legumain/Asparaginyl Endopeptidase in ELISAs and Western blots.
Human
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LGMN
NS0-derived recombinant human Legumain Val18-Tyr433 Accession # Q99538
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5641
Reconstitute at 0.5 mg/mL in sterile PBS.