Learn More
Latent TGF-beta bp2/LTBP-2 Mouse anti-Human, Clone: 327318, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3850
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450286
  • £277.83 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Latent TGF-beta bp2/LTBP-2 Monoclonal antibody MAB3850 Specifically detects Latent TGF-beta bp2/LTBP-2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Latent TGF-beta bp2/LTBP-2
327318
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
LTBP2
Mouse myeloma cell line NS0-derived recombinant human Latent TGF-beta bp2/LTBP-2 Gln36-Glu1821 Accession # NP_000419
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4053
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_000419
C14orf141, GLC3D, Latent TGFbeta bp2, latent transforming growth factor beta binding protein 2, latent-transforming growth factor beta-binding protein 2, LTBP2, LTBP-2, LTBP3chromosome 14 open reading frame 141, MSTP031
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Latent TGF-beta bp2/LTBP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse LTMP-4 is observed.