missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC43A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538543
Code : NEW
Additional Details : Weight : 0.00900kg
Description
SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids. SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids.Specifications
SLC43A1 | |
Polyclonal | |
Unconjugated | |
SLC43A1 | |
Large neutral amino acids transporter small subunit 3, L-type amino acid transporter 3, Prostate cancer overexpressed gene 1 protein, Solute carrier family 43 member 1, LAT3, PB39, POV1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8501 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75387 | |
SLC43A1 | |
A synthetic peptide derived from the internal region of human LAT3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |