missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LASP1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518828
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
LASP1 Polyclonal antibody specifically detects LASP1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
LASP1 | |
Polyclonal | |
Unconjugated | |
Lasp1 | |
AA408629; Def-4; Lasp1; LASP-1; lasp1 {ECO:0000312; LIM and SH3 domain protein 1; LIM and SH3 protein 1; metastatic lymph node gene 50 protein; MLN 50; MLN50; RGD:68408}; SH3P6; Tg(11Col1a1)NG; Tg(Col1a1-lacZ)1Ngma | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3927 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q14847 | |
Lasp1 | |
Synthetic peptide sequence (PERRDSQDGSSYR) corresponding to the internal amino acids of LASP1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |