missing translation for 'onlineSavingsMsg'
Learn More

LAP (TGF-beta 1) Mouse anti-Human, Clone: 27232, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2463

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459465

  • £353.77 / 500µg


Estimated Shipment
03-12-2018
Add to basket

Description

Description

LAP (TGF-beta 1) Monoclonal antibody MAB2463 Specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

LAP (TGF-beta 1)
27232
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P01137
CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human LAP (TGF-beta 1) in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) TGF-beta 1, rhTGF-beta 2, rhTGF-beta 1.2, rhTGF-beta 3, or rhTGF-alpha .
Human
Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TGFB1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1), Leu30-Arg278 (Cys33Ser), Accession # P01137
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7040
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only