Learn More
L3MBTL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530499
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa nucleus, human L3MBTL-transfected 293T. Predicted reactivity: Mouse (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
L3MBTL1 Polyclonal antibody specifically detects L3MBTL1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
L3MBTL1 | |
Polyclonal | |
Unconjugated | |
L3MBTL1 | |
C630004G01; dJ138B7.3; H-l(3)mbt; H-l(3)mbt protein; Kiaa0681; L(3)mbt protein homolog; l(3)mbt-like (Drosophila); l(3)mbt-like 1; l(3)mbt-like 1 (Drosophila); L3mbt; L3MBTL; L3MBTL histone methyl-lysine binding protein 1; L3MBTL1; L3MBTL1, histone methyl-lysine binding protein; lethal (3) malignant brain tumor l(3); lethal(3)malignant brain tumor-like protein 1; lethal(3)malignant brain tumor-like protein 1; serine/threonine-protein kinase Sgk2; LOC100070683; mKIAA0681; RGD1307316; RP1-138B7.1; ZC2HC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26013 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y468 | |
L3MBTL1 | |
Recombinant fragment corresponding to a region within amino acids 82 and 298 of Human L3MBTL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |