Learn More
L-Selectin/CD62L Rat anti-Mouse, Clone: 95218, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5761
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441066
  • £364.33 / 500µg
Estimated Shipment
01-07-2019
Description
Description
L-Selectin/CD62L Monoclonal antibody MAB5761 Specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
L-Selectin/CD62L
95218
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
P18337
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse L-Selectin/CD62L in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human L-Selectin, recombinant mouse (rm) E-Selectin, or rmP-Selectin is observed.
Mouse
Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SELL
Mouse myeloma cell line NS0-derived recombinant mouse L-Selectin/CD62L Met1-Asn332 Accession # P18337
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6402
Reconstitute at 0.5 mg/mL in sterile PBS.