Learn More
L-Selectin/CD62L Mouse anti-Rat, Clone: 196021, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1534
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438715
  • £337.93 / 500µg
Estimated Shipment
08-07-2019
Description
Description
L-Selectin/CD62L Monoclonal antibody MAB1534 Specifically detects L-Selectin/CD62L in Rat samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
L-Selectin/CD62L
196021
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SELL
Mouse myeloma cell line NS0-derived recombinant rat L-Selectin/CD62L Trp39-Asn332 Accession # P30836
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6402
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, ELISA
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P30836
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat L-Selectin/CD62L in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) L-Selectin or rhL-Selectin.
Rat