missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRTAP11-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531957
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KRTAP11-1 Polyclonal antibody specifically detects KRTAP11-1 in Human samples. It is validated for Western Blot.Specifications
KRTAP11-1 | |
Polyclonal | |
Unconjugated | |
KRTAP11-1 | |
AA162070; AA589467; AV018433; Hacl1; HACL-1; High sulfur keratin-associated protein 11.1; KAP11.1; keratin associated protein 11-1; keratin-associated protein 11.1; keratin-associated protein 11-1; KRTAP11.1; KRTAP11-1; PRED29 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
337880 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IUC1 | |
KRTAP11-1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 163 of Human KRTAP11-1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |