missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 26 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546514
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: IDIYCNLLDG EERKSKSTCY KSKGYRPVNS GNQAKDSTEE TIVKTVVEEL Sequence homology: Human: 100%; Rabbit: 77%
Cytokeratin 26 Polyclonal antibody specifically detects Cytokeratin 26 in Human samples. It is validated for Western Blot.Specifications
Cytokeratin 26 | |
Polyclonal | |
Unconjugated | |
KRT26 | |
CK26; CK-26; Cytokeratin-26; K25; K25B; K25IRS2; K26; keratin 25B; keratin 26; keratin 26, type I; keratin, type I cytoskeletal 26; Keratin-25B; keratin-26; KRT25B; KRT26; type I inner root sheath specific keratin 25 irs2; type I inner root sheath-specific keratin-K25irs2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
353288 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q7Z3Y9 | |
KRT26 | |
Synthetic peptide directed towards the C-terminal of human KRT26 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |