missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KPNA1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518236
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
KPNA1 Polyclonal antibody specifically detects KPNA1 in Human samples. It is validated for Western Blot.Specifications
KPNA1 | |
Polyclonal | |
Unconjugated | |
KPNA1 | |
AW494490; IMA2; Importin alpha 2; importin alpha 5; Importin alpha P1; importin alpha-5; importin alpha-S1; importin subunit alpha-1; importin subunit alpha-5; Importin subunit alpha-5, N-terminally processed; importin-alpha-S1; IPOA 1; IPOA1; IPOA5; karyopherin (importin) alpha 1; Karyopherin A2; karyopherin alpha 1; karyopherin alpha 1 (importin alpha 5); karyopherin subunit alpha 1; karyopherin subunit alpha-1; karyopherin/importin alpha-1; KPNA1; KPNA2; m-importin-alpha-S1; mSRP1; NPI1; NPI-1; Nucleoprotein interactor 1; RAG cohort 1; RAG cohort protein 2; RCH 1; RCH1; RCH2; recombination activating gene cohort 2; SRP 1; SRP1; SRP1 alpha; SRP1-beta | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3836 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P52294 | |
KPNA1 | |
Synthetic peptide sequence (TTPGKENFRLKSY) corresponding to the N-terminus amino acids of KPNA1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |