Learn More
KOR Mouse anti-Human, Clone: 387301, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3895
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450306
  • £302.82 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
KOR Monoclonal antibody MAB3895 Specifically detects KOR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
Specifications
KOR
387301
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
OPRK1
NS0 mouse myeloma cell line transfected with human KOR Met1-Val380 Accession # P41145
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4986
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, Immunocytochemistry, CyTOF
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
P41145
kappa opioid receptor, kappa-type opioid receptor, KOR, K-OR-1, KOR-1, Opiate receptor, kappa-1, opioid receptor, kappa 1, OPRK, OPRK1, Oprk2, R21
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human KOR. Stains human KOR transfectants but not irrelevant transfectants.