missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521696
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLRG1 Polyclonal antibody specifically detects KLRG1 in Human samples. It is validated for Western Blot.Specifications
KLRG1 | |
Polyclonal | |
Unconjugated | |
Klrg1 | |
2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10219 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96E93 | |
Klrg1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 195 of KLRG1 (Uniprot ID#Q96E93) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |