Learn More
KLHL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530441
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (98%), Chicken (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KLHL2 Polyclonal antibody specifically detects KLHL2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
KLHL2 | |
Polyclonal | |
Unconjugated | |
KLHL2 | |
6030411N21Rik; ABP-KELCH; actin-binding protein Mayven; AU020744; kelch; kelch like family member 2; kelch-like 2, Mayven; kelch-like family member 2; kelch-like protein 2; KLHL2; MAV; MAYVEN; si:ch211-287c11.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95198 | |
KLHL2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 235 of Human KLHL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |