missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542160
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: IMFEHFFLHK EWISKDPSRL VGQIHDGVSK ANFSIGSMMR ALAGTYRCYG Sequence homology: Human: 100%
KIR3DS1 Polyclonal antibody specifically detects KIR3DS1 in Human samples. It is validated for Western Blot.Specifications
KIR3DS1 | |
Polyclonal | |
Unconjugated | |
KIR3DS1 | |
CD158E2; killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1; killer cell immunoglobulin-like receptor 3DS1; killer cell immunoglobulin-like receptor KIR3DS1; killer cell immunoglobulin-like receptor, three domains, short cytoplasmic tail, 1; killer Ig receptor; KIR-123FM; KIR3DS1; KIR-G1; MHC class I NK cell receptor; natural killer cell receptor; natural killer-associated transcript 10; NKAT10; NKAT-10 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3813 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14943 | |
KIR3DS1 | |
Synthetic peptide directed towards the middle region of human KIR3DS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |