Learn More
KIR3DL1 Mouse anti-Human, Clone: 177407, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB12251
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448465
  • £420.32 / 500µg
Estimated Shipment
17-08-2020
Description
Description
KIR3DL1 Monoclonal antibody MAB12251 Specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
KIR3DL1
177407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KIR3DL1
BaF3 mouse pro-B cell line transfected with human KIR3DL1 Accession # P43629
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3811
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P43629
AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human KIR3DL1 on transfected BaF/3 cells. When tested against a panel of transfectants, no cross-reactivity with KIR2DL1, 2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL2, or 3DS1 is observed.