Learn More
Kininogen Light Chain Rat anti-Mouse, Clone: 288424, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB22061
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439265
  • £311.64 / 100µg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
Kininogen Light Chain Monoclonal antibody MAB22061 Specifically detects Kininogen Light Chain in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Kininogen
288424
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Kininogen Light Chain in direct ELISAs and Western blots. In Western blots, reacts with full length Kininogen (aa 21-661) and several forms of the light chain starting at residue 389, and does not react with the heavy chain. In Western blots, no cross-reactivity with recombinant mouse (rm) Cystatin A, B, C, E/M, recombinant human (rh) Cystatin D, F, S, SA, SN, rmCystatin/Stefin Homolog, rmFetuin A, rhFetuin B, rmHPRG, rhKininogen, or rhKininostatin is observed.
Mouse
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
O08677
Kng1
Mouse myeloma cell line NS0-derived recombinant mouse Kininogen Glu21-Ser661 Accession # O08677
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3827
Reconstitute at 0.5 mg/mL in sterile PBS.