Learn More
Kininogen Heavy Chain Mouse anti-Human, Clone: 236012, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB15692
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438745
  • £337.93 / 500µg
Estimated Shipment date
from Supplier 24-09-2019
Description
Description
Kininogen Heavy Chain Monoclonal antibody MAB15692 Specifically detects Kininogen Heavy Chain in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Kininogen
236012
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Kininogen Heavy Chain in direct ELISAs and Western blots. It reacts with HK, Kininogen, and the heavy chain, but not with the light chain alone. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cystatin A, B, C, D, E, S, SA, SN, rhFetuin A, rhFetuin B, rhHPRG, recombinant mouse Kininogen, or rhKininostatin is observed.
Human
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P01042
KNG1
Mouse myeloma cell line NS0-derived recombinant human Kininogen Gln19-Ser644 (Ile581Thr) Accession # P01042
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3827
Reconstitute at 0.5 mg/mL in sterile PBS.