Learn More
KIN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540419
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: LKTIGSSASV KRKESSQSST QSKEKKKKKS ALDEIMEIEE EKKRTARTDY Sequence homology: Cow: 85%; Dog: 85%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Rabbit: 85%; Rat: 85%
KIN Polyclonal antibody specifically detects KIN in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
KIN | |
Polyclonal | |
Unconjugated | |
Kin | |
antigenic determinant of rec-A protein; antigenic determinant of rec-A protein homolog; binding to curved DNA; Btcd; DNA/RNA-binding protein KIN17; HsKin17 protein; KIN; KIN, antigenic determinant of recA protein; KIN, antigenic determinant of recA protein homolog; KIN17; Kin17 DNA and RNA binding protein; Rts2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22944 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60870 | |
Kin | |
Synthetic peptide directed towards the middle region of human KIN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |