missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540813
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: VRPPSPDGST SQEESPSHFT AVPGEPLGDE TQGQQPLQLE EDQRAWQRLE Sequence homology: Cow: 86%; Horse: 86%; Human: 100%; Mouse: 79%
KIFC2 Polyclonal antibody specifically detects KIFC2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
KIFC2 | |
Polyclonal | |
Unconjugated | |
Kifc2 | |
KIFC2; kinesin family member C2; kinesin-like protein KIFC2 | |
Rabbit | |
Protein A | |
RUO | |
90990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96AC6 | |
Kifc2 | |
Synthetic peptide directed towards the N-terminal of human KIFC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |