missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF18A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531477
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, HCT116. Predicted reactivity: Mouse (88%), Chicken (80%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KIF18A Polyclonal antibody specifically detects KIF18A in Human samples. It is validated for Western Blot.Specifications
KIF18A | |
Polyclonal | |
Unconjugated | |
KIF18A | |
AU024633; B130001M12Rik; Kif18a; kinesin family member 18A; kinesin superfamily protein 18A; kinesin-like protein KIF18A; Marrow stromal KIF18A; MS-KIF18A; N-8 kinesin; OK/SW-cl.108; PPP1R99; protein phosphatase 1, regulatory subunit 99 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NI77 | |
KIF18A | |
Recombinant fragment corresponding to a region within amino acids 1 and 269 of Human KIF18A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |