Learn More
TOX4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541558
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MEFPGGNDNY LTITGPSHPF LSGAETFHTP SLGDEEFEIP PISLDSDPSL Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
TOX4 Polyclonal antibody specifically detects TOX4 in Human samples. It is validated for Western Blot.Specifications
TOX4 | |
Polyclonal | |
Unconjugated | |
TOX4 | |
5730589K01Rik; A630040M18; AA410149; C14orf92; Epidermal Langerhans cell protein LCP1; KIAA0737; LCP1; MIG7; migration-inducing protein 7; RGD708449; TOX high mobility group box family member 4; TOX4; Unknown (protein for MGC:139878) | |
Rabbit | |
Protein A | |
RUO | |
9878 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O94842 | |
TOX4 | |
Synthetic peptide directed towards the N-terminal of human KIAA0737 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |