Learn More
KHK Monoclonal Antibody (S1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA519238
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MEEKQILCVG LVVLDVISLV DKYPKEDSEI RCLSQRWQRG GNASNSCTIL SLLGAPCAFM GSMAPGHVAD FVLDDLRRYS VDLRYTVFQT TGSVPIATVI INEASGSRTI LYYDRSLPDV SATDFEKVDL TQFKWIHIEG RNASEQVKML QRIDAHNTRQ PPEQKIRVSV EVEKPREELF QLFGYGDVVF VSKDVAKHLG FQSAEEALRG LYGRVRKGAV LVCAWAEEGA DALGPDGKLL HSDAFPPPRV VDTLGAGDTF NASVIFSLSQ GRSVQEALRF GCQVAGKKCG LQGFDGIV
KHK Monoclonal antibody specifically detects KHK in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
KHK | |
Monoclonal | |
0.28 mg/mL | |
PBS with no preservative; pH 7.4 | |
P50053 | |
KHK | |
KHK (AAH06233, 1 a.a. ∽ 298 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Western Blot | |
S1 | |
Unconjugated | |
KHK | |
fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102 | |
Mouse | |
Affinity chromatography | |
RUO | |
3795 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |