Learn More
Kell Rat anti-Mouse, Clone: 214711, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1454
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438655
  • £390.72 / 500µg
Estimated Shipment
13-08-2020
Description
Description
Kell Monoclonal antibody MAB1454 Specifically detects Kell in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Kell
214711
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KEL
Mouse myeloma cell line NS0-derived recombinant mouse Kell Phe50-Trp713 Accession # Q9EQF2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3792
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9EQF2
CD238, CD238 antigen, EC 3.4.24.-, ECE3, KEL, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Kell in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) ECE-2, rhNeprilysin, or recombinant mouse Neprilysin is observed.