Learn More
KEAP1 Monoclonal Antibody (1F10B6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517106
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17106 targets KEAP1 in indirect ELISA, FACS, ICC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17106 immunogen is purified recombinant fragment of human KEAP1 (amino acids: 380-624) expressed in E. Coli. MA5-17106 detects KEAP1 which has a predicted molecular weight of approximately 69.7kDa.
KEAP1 Monoclonal antibody specifically detects KEAP1 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
KEAP1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q14145, Q9Z2X8 | |
Keap1 | |
Purified recombinant fragment of human KEAP1 (amino acids: 380-624) expressed in E. Coli. | |
RUO | |
50868, 9817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
1F10B6 | |
Unconjugated | |
Keap1 | |
cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |