missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP60 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519523
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat, fruit fly (Drosophila melanogaster) and zebrafish based on sequence homology.
TIP60 Polyclonal antibody specifically detects TIP60 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
TIP60 | |
Polyclonal | |
Unconjugated | |
KAT5 | |
60 kDa Tat-interactive protein; AI839539; cPLA(2)-interacting protein; CPLA2; cPLA2 interacting protein; ESA1; histone acetyltransferase HTATIP; Histone acetyltransferase KAT5; HIV-1 Tat interactive protein; HIV-1 tat interactive protein 1, 60 kDa homolog; HIV-1 tat interactive protein 1, homolog; HIV-1 Tat interactive protein 60 kD; HIV-1 Tat interactive protein, 60 kD; HIV-1 Tat interactive protein, 60kDa; HIV-1 tat interactive protein, homolog; HTATIP; Htatip1; K(lysine) acetyltransferase 5; K-acetyltransferase 5; Kat5; Lysine acetyltransferase 5; PLIP; Tat interacting protein, 60kDa; Tat-interactive protein-60; TIP; Tip55; Tip60; TIP60(HTATIP); Tip60b; ZC2HC5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10524, 81601 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
Q8CHK4, Q92993 | |
KAT5 | |
Synthetic peptide conjugated to KLH derived from within residues 450 to the C-terminus of Human Tip60. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |