Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, Biotin, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA512114
Code : LE
Additional Details : Weight : 0.01000kg
Description
By Western blot, MA5-12114 detects Human Ig kappa light chain chain at ∽25kD in human samples. At high concentrations, the antibody will also detect heavy chain. By ELISA and WB, MA5-12114 does not react with human Ig lambda light chain.
Human Kappa Light Chain Monoclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Human Kappa Light Chain | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
L1C1 | |
Biotin | |
Immunoglobulin kappa light chain, Ig kappa light chain, Ig kappa, kappa light chain, Igk | |
B lymphoma cells | |
500 μL | |
Secondary | |
4° C | |
IgG1 |