missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 6, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA535313

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15328034

  • £304.30 / 400µL


Estimated Shipment
05-07-2018
Add to basket

Specifications

Specifications

Kallikrein 6
Polyclonal
PBS with 0.09% sodium azide
Q92876
KLK6, PRSS18, PRSS9, Kallikrein-6, Neurosin, Protease M, SP59, Serine protease 18, Serine protease 9, Zyme
Rabbit
IgG
400μL
Antibody
Polyclonal
Human
Flow CytometryþImmunohistochemistry (Paraffin)þWestern Blot
Unconjugated
KLK6
Purified
KLK6
KLH conjugated synthetic peptide between 126-156 amino acids from the central region of human Kallikrein 6
Ammonium sulfate precipitation, Size-exclusion - Dialysis
Store at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
5653
Videos