Learn More
Kallikrein 6/Neurosin Mouse anti-Human, Clone: 265508, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2008
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459095
  • £249.49 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Kallikrein 6/Neurosin Monoclonal antibody MAB2008 Specifically detects Kallikrein 6/Neurosin in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Kallikrein 6/Neurosin
265508
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KLK6
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 6/Neurosin Leu22-Lys244 Accession # Q92876
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5653
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q92876
Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, KLK6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Kallikrein 6/Neurosin in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Factor X, recombinant mouse HGFA, rhKLK3, 4, 5, 8, 10, 11, 15, rhThrombin, or rhuPA is observed.