missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527208
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Kallikrein 2 Polyclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Kallikrein 2 | |
Polyclonal | |
Unconjugated | |
KLK2 | |
glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P20151 | |
KLK2 | |
Recombinant fragment corresponding to a region within amino acids 112 and 261 of Human Kallikrein 2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |