missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 2 Mouse anti-Human, Clone: 426723, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4104

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460406

  • £264.01 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

Kallikrein 2 Monoclonal antibody MAB4104 Specifically detects Kallikrein 2 in Human samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Specifications

Specifications

Kallikrein 2
426723
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KLK2
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 2, Ile25-Pro261, Accession # P20151
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3817
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation, Neutralization
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3.4 ug/mL
P20151
EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, Klk1c2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2, Ton
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human Kallikrein 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, or B1 is observed.
Human
Videos

For Research Use Only