missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 14 Mouse anti-Human, Clone: 337903, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2626

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449585

  • £277.83 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 14 Monoclonal antibody MAB2626 Specifically detects Kallikrein 14 in Human samples. It is validated for Immunoprecipitation, Neutralization
Specifications

Specifications

Kallikrein 14
337903
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KLK14
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248 Accession # AAD50773
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
43847
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunoprecipitation, Neutralization
LYOPH
Immunoprecipitation 25 ug/mL, Neutralization 0.07 ug/mL
AAD50773
EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Kallikrein 14 in direct ELISAs.
Human
Videos
Documents

For Research Use Only