missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 13 Mouse anti-Human, Clone: 341419, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2625

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439585

  • £337.93 / 500µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Kallikrein 13 Monoclonal antibody MAB2625 Specifically detects Kallikrein 13 in Human samples. It is validated for Immunoprecipitation, Neutralization, Western Blotting
Specifications

Specifications

Kallikrein 13
341419
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.4 ug/mL
Q9UKR3
DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Kallikrein 13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Kallikrein 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 14, 15, rhKallikrein B1, rhFactorVII, rhHGFA, rhUPA, rhThrombin, rhFactor X, or rhFactor XI is observed.
Human
Western Blot, Immunoprecipitation, Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
KLK13
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 13 Val19-Ile262 Accession # Q9UKR3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
26085
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only