missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jouberin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521895
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Jouberin Polyclonal antibody specifically detects Jouberin in Human samples. It is validated for Western Blot.Specifications
Jouberin | |
Polyclonal | |
Unconjugated | |
AHI1 | |
1700015F03Rik; Abelson helper integration site 1; abelson helper integration site 1 protein; abelson helper integration site 1 protein homolog; Ahi1; AHI-1; contatins SH3 and WD40 domains; D10Bwg0629e; dJ71N10.1; JBTS3; jouberin; ORF1; RP1-32B1.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8N157 | |
AHI1 | |
Synthetic peptide corresponding to a region within amino acids 1 and 42 of AHI1 (Uniprot ID#Q8N157) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |