missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535012
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
JMJD3 Polyclonal antibody specifically detects JMJD3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
JMJD3 | |
Polyclonal | |
Unconjugated | |
KDM6B | |
1700064E03Rik; BC038313; jmjC domain-containing protein 3; JMJD3; jumonji domain containing 3; jumonji domain containing 3, histone lysine demethylase; jumonji domain-containing 3; jumonji domain-containing protein 3; KDM1 lysine (K)-specific demethylase 6B; Kdm6b; KIAA0346; lysine (K)-specific demethylase 6B; lysine demethylase 6B; Lysine-specific demethylase 6B; mKIAA0346 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
23135 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
O15054 | |
KDM6B | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human JMJD3 | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |