missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA511148
Code : LE
Additional Details : Weight : 0.01000kg
Description
JMJD2D Polyclonal antibody specifically detects JMJD2D in Human samples. It is validated for Western Blot.Specifications
JMJD2D | |
Polyclonal | |
Unconjugated | |
KDM4D | |
FLJ10251; JHDM3D; JmjC domain-containing histone demethylation protein 3D; JMJD2D; jumonji domain containing 2D; jumonji domain-containing protein 2D; KDM4D; lysine (K)-specific demethylase 4D; lysine demethylase 4D; lysine-specific demethylase 4D | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
55693 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.8 mg/mL | |
PBS with 0.09% sodium azide | |
Q6B0I6 | |
KDM4D | |
KLH conjugated synthetic peptide between 484-516 amino acids from the C-terminal region of human JMJD2D | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |