Learn More
JAM-A Rat anti-Mouse, Clone: 160407, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1077
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428315
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
JAM-A Monoclonal antibody MAB1077 Specifically detects JAM-A in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
JAM-A
160407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
F11R
Mouse myeloma cell line NS0-derived recombinant mouse JAM-A Lys27-Ala242 Accession # O88792
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
50848
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O88792
CD321, CD321 antigen, F11 receptor, F11R, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1, PAM-1KAT, Platelet adhesion molecule 1
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse JAM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human JAM-A.