missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44406G
Code : 50
Additional Details : Weight : 0.50000kg
Description
JAK2 Polyclonal antibody specifically detects JAK2 in Human, Mouse samples. It is validated for Western Blot.Specifications
JAK2 | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
O60674, Q62120 | |
JAK2 | |
The antiserum was produced against a chemically synthesized peptide derived from the C-terminal region of human JAK2. The sequence is conserved in mouse and rat. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
JAK2 | |
EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16452, 3717 | |
-20°C | |
Liquid |