missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged2 Monoclonal Antibody (T.584.4), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514978
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Jagged1.
Jagged2 Monoclonal antibody specifically detects Jagged2 in Human, Rat samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Jagged2 | |
Monoclonal | |
153 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P97607, Q9Y219 | |
Jag2 | |
Synthetic peptide corresponding to residues surrounding Ala117 of human Jagged2 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
T.584.4 | |
Unconjugated | |
Jag2 | |
D12Ggc2e; HJ2; JAG2; jagged 2; Jagged2; mJagged2-1; protein jagged-2; SER2; Serh; sm; syndactylism | |
Rabbit | |
Affinity chromatography | |
RUO | |
29147, 3714 | |
-20°C | |
Liquid |