missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49f Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA512334
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken based on sequence homology.
CD49f Polyclonal antibody specifically detects CD49f in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD49f | |
Polyclonal | |
Unconjugated | |
Itga6 | |
5033401O05Rik; AI115430; Alpha6p; CD49 antigen-like family member F; CD49f; DKFZp686J01244; FLJ18737; integrin alpha 6; integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin subunit alpha 6; integrin, alpha 6; Itga6; ITGA6B; LOW QUALITY PROTEIN: integrin alpha-6; Processed integrin alpha-6; VLA-6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3655 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P23229 | |
Itga6 | |
KLH conjugated synthetic peptide between 1043-1072 amino acids from human ITGA6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |