missing translation for 'onlineSavingsMsg'
Learn More

IREB2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA116543

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11525313

  • £332.00 / 100┬ÁL


Estimated Shipment
20-06-2018
Add to basket

Description

Description

This antibody is predicted to work in human based on 100% sequence homology, however preliminary data shows poor reactivity against human protein.This antibody also has 72% sequence homology with rat.Suggested positive control: 3T3 whole cell lysate.
Specifications

Specifications

IRP2
Polyclonal
Unconjugated
IREB2
Purified
IREB2
Synthetic peptide corresponding to the N-terminal residues 1-100 of human IRP2 (UniProt P48200).
Antigen affinity chromatography
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
3658
Western Blot
1mg/mL
tris glycine with 0.15M NaCl and 0.05% sodium azide
P48200, Q811J3
Iron-responsive element binding protein 2, IRE-BP 2, Iron regulatory protein 2
Rabbit
IgG
100μL
Antibody
Polyclonal
Murine
Videos