Learn More
IRAK4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529700
Code : PN
Additional Details : Weight : 0.01000kg
Description
PA5-29700 targets IRAK4 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-29700 immunogen is recombit fragment corresponding to a region within amino acids 27 and 288 of Human IRAK4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes a kinase that activates NF-kappaB in both the Toll-like receptor (TLR) and T-cell receptor (TCR) signaling pathways. The protein is essential for most innate immune responses. Mutations in this gene result in IRAK4 deficiency and recurrent invasive pneumococcal disease. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
IRAK4 | |
Polyclonal | |
Unconjugated | |
Q9NWZ3 | |
IRAK4 | |
Recombit fragment corresponding to a region within amino acids 27 and 288 of Human IRAK4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.76 mg/mL | |
IRAK4 | |
IPD1, IRAK-4, NY-REN-64, REN64, renal carcinoma antigen NY-REN-64 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51135 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |