Learn More
NLRC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520014
Code : LD
Additional Details : Weight : 0.01000kg
Description
In Western blot applications, this antibody detects a band at ∽110kDa. A suggested positive control is human peripheral blood leukocyte (pbl) lysate. PA5-20014 can be used with blocking peptide PEP-0133.
NLRC4 Polyclonal antibody specifically detects NLRC4 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
NLRC4 | |
Polyclonal | |
Unconjugated | |
Nlrc4 | |
9530011P19Rik; AIFEC; CARD, LRR, and NACHT-containing protein; Card12; caspase recruitment domain family, member 12; caspase recruitment domain-containing protein 12; CLAN; Clan protein; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; ice protease-activating factor; ICE-protease activating factor; ipaf; NLR family CARD domain containing 4; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NLRC4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58484 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9NPP4 | |
Nlrc4 | |
A synthetic peptide corresponding to amino acids near the C-terminus of human Ipaf. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |