Learn More
INTS4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530733
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (96%), Rat (95%), Xenopus laevis (85%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
INTS4 Polyclonal antibody specifically detects INTS4 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
INTS4 | |
Polyclonal | |
Unconjugated | |
INTS4 | |
2610034N24Rik; AA692684; AW493223; int4; Integrator complex subunit 4; Ints4; MST093; MSTP093; RGD1305036 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101861, 92105 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.6 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8CIM8, Q96HW7 | |
INTS4 | |
Recombinant fragment corresponding to a region within amino acids 1 and 180 of Human INTS4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |