Learn More
anti-Integrin αVβ3 (Vitronectin Receptor; CD51/CD61), Clone: LM609, Millipore™ (Chemicon™)
Brand:  Merck Millipore MAB1976Z
Code : LD
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10628734
  • £460.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Please note: We no longer supply Millipore Antibodies.
Description
Description
Cheresh, 1987.
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Specifications
Specifications
Integrin αVβ3 (Vitronectin Receptor; CD51/CD61)
Semi-purified
Purified adhesion receptor from the human melanoma cell line M21
100μg
Cell Structure
Monoclonal
Unconjugated
Murine
IgG1
RUO
Primary
AvianþBovineþCanineþChickenþHumanþMonkeyþPorcineþRabbit
Videos