missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 7 Rat anti-Mouse, Clone: 332408, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB30601

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439865

  • £288.60 / 100µg

Estimated Shipment
18-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Integrin beta 7 Monoclonal antibody MAB30601 Specifically detects Integrin beta 7 in Mouse samples. It is validated for Adhesion Activation
Specifications

Specifications

Integrin beta 7
332408
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ITGB7
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin beta 7 Glu20-His722 Accession # P26011
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3695
Reconstitute at 0.5 mg/mL in sterile PBS.
Adhesion Activation
LYOPH
Adhesion Activation
P26011
Gut homing receptor beta subunit, integrin beta 7 subunit, integrin beta-7, integrin, beta 7, ITGB7
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Integrin beta 7 extracellular domain in direct ELISAs.
Mouse
Videos
Documents

For Research Use Only