missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha V beta 3 Mouse anti-Human, Clone: 23C6, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB3050

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13469855

  • £320.46 / 100µg

Estimated Shipment date
from Supplier 25-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Integrin alpha V beta 3 Monoclonal antibody MAB3050 Specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Blocking (Adhesion), Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Specifications

Specifications

Integrin alpha V beta 3
23C6
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
antigen identified by monoclonal L230, CD51, CD51 antigen, CD61, GP3A, GPIIIA, INGRB3, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), ITGAV, ITGB3, MSK8, Vitronectin Receptor alpha, Vitronectin receptor subunit alpha, VNRA, VNRADKFZp686A08142
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Integrin alpha V beta 3.
Human
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Adhesion Blockade, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, Adhesion Blockade, CyTOF-ready
Purified
ITGAV
Human osteoclasts
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3685
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only