missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA5 Monoclonal Antibody (238307), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA523585
Code : NEW
Additional Details : Weight : 0.00010kg
Description
ITGA5 Monoclonal antibody specifically detects ITGA5 in Human samples. It is validated for Flow Cytometry.Specifications
ITGA5 | |
Monoclonal | |
APC | |
Itga5 | |
2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A | |
Mouse | |
Protein A/G | |
RUO | |
3678 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
238307 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P08648 | |
Itga5 | |
Mouse myeloma cell line NS0-derived recombinant human Integrin alpha 5/CD49e Phe42-Tyr995 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |