Learn More
Integrin alpha 4/CD49d Rat anti-Mouse, Clone: 265329, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2450
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439455
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Integrin alpha 4/CD49d Monoclonal antibody MAB2450 Specifically detects Integrin alpha 4/CD49d in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
Integrin alpha 4/CD49d
265329
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ITGA4
Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 4/CD49d Tyr34-Phe976 Accession # Q792F9
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3676
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q792F9
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), ITGA4, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4, VLA-4 subunit alpha
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Integrin alpha 4/CD49d.